missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) oxide pellets, 99.9% (REO), Thermo Scientific Chemicals

Sintered pellets suitable for vacuum deposition

582.00 € - 1701.00 €

Chemical Identifiers

CAS 1314-36-9
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11348578
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040648.22
100 g
582.00 €
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11358578
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040648.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Sintered pellets suitable for vacuum depositionYttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is a precursor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-36-9
225.81
RUDFQVOCFDJEEF-UHFFFAOYSA-N
[O--].[O--].[O--].[Y+3].[Y+3]
O3Y2
MFCD00011473
diyttrium(3+) trioxidandiide

Specifications

2410°C
O3Y2
14,10107
RUDFQVOCFDJEEF-UHFFFAOYSA-N
diyttrium(3+) trioxidandiide
225.81
Odorless
5.01 g/mL
Yttrium(III) oxide pellets, approx. 10mm dia. x 5mm thick
1314-36-9
MFCD00011473
Insoluble in water. Soluble in alcohol and dilute acids.
[O--].[O--].[O--].[Y+3].[Y+3]
225.81
99.9%
Air Sensitive
(REO)
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 215-233-5

missing translation for 'rtecsNumber' : ZG3850000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only