Learn More
Yttrium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
123.00 € - 540.00 €
Chemical Identifiers
CAS | 1314-36-9 |
---|---|
Molecular Formula | O3Y2 |
Molecular Weight (g/mol) | 225.81 |
MDL Number | MFCD00011473 |
InChI Key | RUDFQVOCFDJEEF-UHFFFAOYSA-N |
IUPAC Name | diyttrium(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Y+3].[Y+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11326198
|
Thermo Scientific Alfa Aesar
011181.22 |
100 g |
123.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336198
|
Thermo Scientific Alfa Aesar
011181.A1 |
1 kg |
540.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Yttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is presurcor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1314-36-9 | |
225.81 | |
RUDFQVOCFDJEEF-UHFFFAOYSA-N | |
[O--].[O--].[O--].[Y+3].[Y+3] |
O3Y2 | |
MFCD00011473 | |
diyttrium(3+) trioxidandiide |
Specifications
2410°C | |
Powder | |
O3Y2 | |
Insoluble in water. Soluble in alcohol and dilute acids | |
[O--].[O--].[O--].[Y+3].[Y+3] | |
225.81 | |
5.01 g/mL |
1314-36-9 | |
99.99% (REO) | |
MFCD00011473 | |
RUDFQVOCFDJEEF-UHFFFAOYSA-N | |
diyttrium(3+) trioxidandiide | |
225.81 | |
Yttrium(III) oxide |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 215-233-5
missing translation for 'rtecsNumber' : ZG3850000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only