missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) oxide, REacton™, 99.999% (REO), Thermo Scientific Chemicals

80.20 € - 636.17 €

Chemical Identifiers

CAS 1314-36-9
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11366198
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011182.14
25 g
80.20 €
25g
Please to purchase this item. Need a web account? Register with us today!
11356198
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011182.22
100 g
231.15 €
100g
Please to purchase this item. Need a web account? Register with us today!
11376198
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011182.A1
1 kg
636.17 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Yttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is presurcor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-36-9
225.81
RUDFQVOCFDJEEF-UHFFFAOYSA-N
[O--].[O--].[O--].[Y+3].[Y+3]
O3Y2
MFCD00011473
diyttrium(3+) trioxidandiide

Specifications

2410°C
White
99.999% (REO)
MFCD00011473
RUDFQVOCFDJEEF-UHFFFAOYSA-N
diyttrium(3+) trioxidandiide
225.81
5.01
1314-36-9
Powder
O3Y2
Insoluble in water. Soluble in alcohol dilute acids
[O--].[O--].[O--].[Y+3].[Y+3]
225.81
Odorless
Yttrium(III) oxide
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 215-233-5

missing translation for 'rtecsNumber' : ZG3850000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only