missing translation for 'onlineSavingsMsg'
Learn More
Yttrium(III) phosphate hydrate, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11304997

Yttrium(III) phosphate hydrate, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11304997
10 g
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy

Product Code. 11304997

Brand: Thermo Scientific Alfa Aesar 041991.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The most important use of yttrium is in making phosphors, particularly the red phosphors used in television set cathode ray tube (CRT) displays and in LEDs. It is also used in the production of electrodes, electrolytes, electronic filters, lasers and superconductors; various medical applications; and the tracing of various materials to enhance their properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34054-55-2
Molecular Formula O4PY
Molecular Weight (g/mol) 183.88
MDL Number MFCD00049782
InChI Key UXBZSSBXGPYSIL-UHFFFAOYSA-K
Synonym yttrium iii phosphate hydrate, acmc-20aj99, yttrium phosphate-water 1/1/1, yttrium 3+ hydrate phosphate, yttrium 3+ ion hydrate phosphate
PubChem CID 57372096
IUPAC Name yttrium(3+) phosphate
SMILES [Y+3].[O-]P([O-])([O-])=O

Specifications

Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 183.88 (Anhydrous)
Percent Purity 99.99%
Assay (REO)
Chemical Name or Material Yttrium(III) phosphate hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberF08L06 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041991.09,041991.04
Lot NumberL15K29 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041991.09,041991.04
Lot NumberH08L19 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number041991.09,041991.04
Lot NumberJ05L18 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number041991.09,041991.04
Lot NumberL30M03 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number041991.09,041991.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium(III) phosphate hydrate, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.