missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) sulfate octahydrate, REacton™, 99.9% (REO)

Product Code. p-7049504
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11332508

Brand: Thermo Scientific Alfa Aesar 014561.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7446-33-5 | H16O20S3Y2 | 610.10 g/mol

Yttrium(III) sulfate octahydrate is used as a laboratory reagent, colorant, structural ceramics, optical glasses and catalysts. It finds application in electrical components and photo-optical materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-33-5
Molecular Formula H16O20S3Y2
Molecular Weight (g/mol) 610.10
MDL Number MFCD00149946
InChI Key OMBXBIHFXZAMLA-UHFFFAOYSA-H
Synonym yttrium iii sulfate octahydrate, yttrium sulfate octahydrate, sulfuric acid, yttrium 3+ salt 3:2 , octahydrate, yttrium iii sulfate octahydrate, reacton reo, diyttrium trisulfate octahydrate, ytterium sulphate, yttrium sulfate,, yttrium 3+ trisulfate octahydrate, yttrium iii sulfate octahydrate, reacton, diyttrium 3+ octahydrate trisulfate
PubChem CID 165608
IUPAC Name yttrium(3+);trisulfate;octahydrate
SMILES O.O.O.O.O.O.O.O.[Y+3].[Y+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color White to Yellow
Physical Form Crystalline
Quantity 50 g
Merck Index 14,10107
Solubility Information Water solubility decreases with increasing temperature; Soluble in water.
Formula Weight 610.11 (465.99 Anhydrous)
Percent Purity 99.9%
Grade Reacton
Odor Odorless
Assay (REO)
Chemical Name or Material Yttrium(III) sulfate octahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.