missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) sulfide, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11388378
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12039-19-9
Molecular Formula S3Y2
Molecular Weight (g/mol) 273.99
MDL Number MFCD00058806
InChI Key BPMRLDMEAPSVQN-UHFFFAOYSA-N
Synonym yttrium sulfide, diyttrium trisulphide, yttrium iii sulfide, yttrium sulfide y2s3, yttrium 3+ trisulfide, diyttrium 3+ ion trisulfanediide, yttrium iii sulfide trace rare earth metals basis 1g
PubChem CID 166021
SMILES [S--].[S--].[S--].[Y+3].[Y+3]

Specifications

Physical Form Powder
Quantity 5 g
UN Number UN3134
Solubility Information Soluble in water.
IUPAC Name yttrium(3+);trisulfide
Formula Weight 273.93
Percent Purity 99.9%
Odor Like rotten eggs
Assay (REO)
Chemical Name or Material Yttrium(III) sulfide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.