missing translation for 'onlineSavingsMsg'
Learn More

Yttrium ingot, 99.9% (REO), Thermo Scientific Chemicals

128.64 € - 486.42 €

Chemical Identifiers

CAS 7440-65-5
Molecular Formula Y
Molecular Weight (g/mol) 88.91
MDL Number MFCD00011468
InChI Key VWQVUPCCIRVNHF-UHFFFAOYSA-N
Synonym yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
PubChem CID 23993
ChEBI CHEBI:33331
IUPAC Name yttrium
SMILES [Y]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11316547
View Documents
Thermo Scientific Alfa Aesar
000399.09
10 g
128.64 €
10g
Please to purchase this item. Need a web account? Register with us today!
11306547
View Documents
Thermo Scientific Alfa Aesar
000399.18
50 g
486.42 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in air stable poly bottle
Specifications

Chemical Identifiers

7440-65-5
88.91
VWQVUPCCIRVNHF-UHFFFAOYSA-N
23993
yttrium
Y
MFCD00011468
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
CHEBI:33331
[Y]

Specifications

7440-65-5
MFCD00011468
VWQVUPCCIRVNHF-UHFFFAOYSA-N
yttrium
23993
99.9%
Odorless
Ingot
Yttrium ingot
Y
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
[Y]
88.91
CHEBI:33331
Contains less than 1% tantalum
Poly bottle
(REO)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-174-8

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Packaged in air stable poly bottle

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.