missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yttrium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals
148.74 € - 626.12 €
Chemical Identifiers
CAS | 7440-65-5 |
---|---|
Molecular Formula | Y |
Molecular Weight (g/mol) | 88.91 |
MDL Number | MFCD00011468 |
InChI Key | VWQVUPCCIRVNHF-UHFFFAOYSA-N |
Synonym | yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element |
PubChem CID | 23993 |
ChEBI | CHEBI:33331 |
IUPAC Name | yttrium |
SMILES | [Y] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11303858
|
Thermo Scientific Alfa Aesar
040299.HF |
10 mm |
148.74 €
10mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11323858
|
Thermo Scientific Alfa Aesar
040299.DH |
25 mm |
261.30 €
25mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11313858
|
Thermo Scientific Alfa Aesar
040299.DJ |
50 mm |
417.08 €
50mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11393848
|
Thermo Scientific Alfa Aesar
040299.DM |
100 mm |
626.12 €
100mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Chemical Identifiers
7440-65-5 | |
88.91 | |
VWQVUPCCIRVNHF-UHFFFAOYSA-N | |
23993 | |
yttrium |
Y | |
MFCD00011468 | |
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element | |
CHEBI:33331 | |
[Y] |
Specifications
7440-65-5 | |
MFCD00011468 | |
VWQVUPCCIRVNHF-UHFFFAOYSA-N | |
yttrium | |
23993 | |
99.9% | |
Mylar under Argon | |
Cast Rod | |
Yttrium rod, 6.35mm (0.25 in.) dia. |
Y | |
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element | |
[Y] | |
88.91 | |
CHEBI:33331 | |
Odorless | |
≈0.14g/mm | |
(metals basis excluding Ta) |
Safety and Handling
missing translation for 'einecsNumber' : 231-174-8
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only