missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 13 Peptide
Z'-LYTE™ Kinase Assay Kit - Ser/Thr 13 Peptide
Brand: Invitrogen™ PV3793
This item is not returnable.
View return policy
Description
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon myosin light chain 2, containing Ser-20 and a Hyd-X-R-X-X-(S⁄T) consensus sequence, where Hyd is a hydrophobic residue and X is any residue.Specifications
Z´-LYTE™ | |
FRET | |
Z´-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions. | |
Methylcoumarin | |
800 x 20 μL Assays | |
myosin light chain 2 (KKRPQRRYSNVF) | |
800 Reactions | |
End Point | |
Peptide-Based Substrate | |
CAMK4, CDC42BPA, CDC42BPB, DAPK3, MYLK2, ROCK2 |
Serine/Threonine Kinases | |
Serine/Threonine Kinase Assay | |
Biochemical phospho-specific activity (no antibody) | |
Kinase Assay Kit | |
384-well plate | |
405⁄465, 535 | |
JBC(2001) 276, p38956 | |
Dry Ice | |
ROCK2, MYLK2, DAPK3, CDC42BPA, CDC42BPB, CAMK4 |
For Research Use Only. Not for use in diagnostic procedures.