Learn More
Thermo Scientific™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 17 Peptide
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format.
Brand:  Thermo Scientific™ PV4316
Product Code. 10328882
  • 812.00 € / 16000µL
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon glycogen synthase, containing Ser-7.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Z'-LYTE™ Ser/Thr 17 Peptide Kinase Assay Kit, 800 x 20μL, 800rxns, Serine/Threonine Kinase Assay Type, Peptide-Based Substrate, 535nm Emission, 405/465nm Excitation, Methylcoumarin Label, End Point Readout, Serine/Threonine Kinases Target Enzyme, FRET Technique, Used for Pharma and Biopharma, Protein Assays and Analysis
Pharma and biopharma, protein biology, drug discovery and development, target and lead identification and validation, kinase biology, biochemical kinase assays, z and acute-lyte™ kinase assays, enzyme and protein activity assays, protein assays and, FRET
800 x 20μL
Biochemical phospho-specific activity (no antibody)
Z'-LYTE™ Ser/Thr 17 Peptide Kinase Assay Kit
384-well plate
Methylcoumarin
End Point
Dry Ice
Peptide-Based Substrate
Serine/Threonine Kinases
Z ́-LYTE™
Z'-LYTE™ Ser/Thr 11 peptide, Z'-LYTE™ Ser/Thr 11 phospho-peptide, 5X kinase buffer A, ATP, development reagent B, development buffer
Serine/Threonine Kinase Assay
–80°C, Z'-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions.
800 x 20μL, 800 x 20 μL
405/465, 535 nm
800 Reactions
Biochem J. vol 296, pp. 843-9 (1993)
Similar to Ser-7 of Glycogen Synthase KKLNRTLSVA (KKLNRTLSFAEPG)
CAMK2B, PRKCN, PRKD2, DCAMKL2, PRKD1, NEK4, MELK
CAMK2B, DCAMKL2 , MELK, NEK4, PRKCN , PRKD1, PRKD2