missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Z'-LYTE™ Kinase Assay Kit - Tyrosine 5 Peptide
Click to view available options
Quantity:
800 x 20μL, 800 x 20 μL Assays
Unit Size:
800 reactions
Description
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon an optimal peptide substrate for ErbB2.
Specifications
Specifications
| Description | Z'-LYTE™ Tyrosine 5 Peptide Kinase Assay Kit, 800 x 20μL, 800rxns, Tyrosine Kinase Assay Type, Peptide-Based Substrate, 535nm Emission, 405/465nm Excitation, Methylcoumarin Label, End Point Readout, Threonine Kinases Target Enzyme, FRET Technique, |
| Product Line | Z ́-LYTE |
| For Use With (Application) | Pharma and biopharma, protein biology, drug discovery and development, target and lead identification and validation, kinase biology, biochemical kinase assays, z and acute-lyte™ kinase assays, enzyme and protein activity assays, protein assays and, FRET |
| Includes | Z'-LYTE™ Ser/Thr 11 peptide, Z'-LYTE™ Ser/Thr 11 phospho-peptide, 5X kinase buffer A, ATP, development reagent B, development buffer |
| Sample Volume | 800 x 20μL |
| Assay | Tyrosine Kinase Assay |
| Assay Entry | Biochemical phospho-specific activity (no antibody) |
| Content And Storage | –80°C, Z'-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions. |
| Product Type | Z'-LYTE™ Tyrosine 5 Peptide Kinase Assay Kit |
| Quantity | 800 x 20μL, 800 x 20 μL Assays |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction