missing translation for 'onlineSavingsMsg'
Learn More

(Z)-Stilbenediboronic acid bis(pinacol) ester, 98%, Thermo Scientific™

Product Code. 11355465
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 221006-76-4
Molecular Formula C26H34B2O4
Molecular Weight (g/mol) 432.174
MDL Number MFCD02683500
InChI Key YFLBNMYBKXYNJN-UHFFFAOYSA-N
Synonym 2-1,2-diphenyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 4377508
IUPAC Name 2-[1,2-diphenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C(=C(B2OC(C(O2)(C)C)(C)C)C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 176°C to 179°C
Quantity 1 g
Beilstein 9081275
Formula Weight 432.18
Percent Purity 98%
Chemical Name or Material (Z)-Stilbenediboronic acid bis(pinacol) ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'