Learn More
LNI Swissgas ZA FID AIR C PLUS 3.0
Zero air generator for FID.
Brand:  LNI Swissgas 6920.28.030
Product Code. 17812019
  • 6710.00 € / Each
Explore more special offers
Description
Description
  • Zero air generator for FID 3.0 L/min.
Specifications
Specifications
100/115/220/230 V
Zero Air Gas Generator
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
LNI Swissgas ZA FID AIR C PLUS 3.0 > LNI Swissgas Zero Air Generator