Learn More
Emma Safety Footwear Zandvoort Safety Shoes
A mid-level boot with water repellant oiled Nubuck uppers, internal steel toe protection and an exceptional sticky grip even on wet and greasy surfaces. Primarily designed for Metal workers, the Zandvoort is used in outdoor occupations where a strong, safe, comfortable shoe is essential.
Brand:  Emma Safety Footwear MM53884648
 View more versions of this product
Product Code. 15908701
  • 170.00 € / 1 pair
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck construction
  • GripForce™ Duo Heat Resistant Outsole
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Specifications
EN ISO 20345:2011
Zandvoort
Oiled Nubuck
Brown
Full grain Nappa leather / Split leather
48