missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zearalanone, Honeywell Fluka™
Click to view available options
Quantity:
5 mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Specifications
Specifications
CAS | 5975-78-0 |
Packaging | Glass Bottle |
Quantity | 5 mg |
PubChem CID | 108003 |
ChEBI | CHEBI:35051 |
Grade | Analytical Standard |
For Use With (Application) | Mycotoxins |
Physical Form | Solid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction