missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zenon™ pHrodo™ iFL IgG Labeling Reagents

Product Code. p-9230008
missing translation for 'orderingAttributeHoverText'
Quantity:
250 μL
1.2 mL
Species:
Human
Mouse
Label or Dye:
pHrodo Deep Red
pHrodo Green
pHrodo Red
pHrodo iFL Green
pHrodo iFL Red
Color:
Deep Red
Green
Red
missing translation for 'unitSize'
1.2mL
240µL
Each
This item is not returnable. View return policy

Product Code. 17964691

missing translation for 'mfr': Invitrogen™ Z25618

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zenon™ pHrodo™ iFL Green-labeled or Red-labeled Fab fragments (i.e. labeling reagent) are directed against the Fc portion of an intact IgG primary antibody to form a labeling complex.

Streamline antibody-drug conjugate screening campaigns with Invitrogen Zenon pHrodo IgG Labeling Reagents to screen for antibody internalization. These reagents provide rapid, scalable antibody labeling and give a bright fluorescence-based indication of antibody uptake and trafficking in target cells when analyzed by flow cytometry, fluorescence microscopy, or high-content screening.

Zenon™ pHrodo™ iFL Green-labeled or Red-labeled Fab fragments (i.e. labeling reagent) are directed against the Fc portion of an intact IgG primary antibody to form a labeling complex. Formation of the Fab–antibody complex occurs in fewer than 5 minutes. Because the pHrodo™ iFL dyes dramatically increase in fluorescence as the pH of their surroundings becomes more acidic, the antibodies coupled with the Zenon™ pHrodo™ iFL IgG Labeling Reagents provide an excellent measure of endocytic activity based on the acidification of the labeled antibodies as they are ingested by the cell.

The Zenon™ pHrodo™ iFL IgG Labeling Reagents contain sufficient material to label one to four 96-well plates, depending on the Zenon™ labeling reagent concentration used. Only a small amount of starting material is needed, and the method is optimized for efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions.

Other advantages of the pH-sensitive Zenon™ pHrodo™ iFL IgG Labeling Reagents include:

  • Simplicity—No reagent purification is required
  • Accuracy—No wash steps or quenchers are needed, minimizing cross-reactivity or background
  • Speed—Labeled antibodies are typically ready to use in 10 minutes
  • Specificity—Dyes are designed for the specific detection of phagocytosis and endocytosis

Preserve primary antibody function and affinities

Reactive dye labeling of primary antibodies can have unpredictable and undesirable outcomes. One of these is reduced binding affinity due to label addition in the binding pocket. The Zenon™ antibody labeling approach, which is targeted to the Fc tail, resolves this potential issue.

Try custom antibody conjugates

If the antibody conjugate you need is not available as one of our stocked products, we will prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

Order Info

Store in refrigerator at 2°C to 8°C.

TRUSTED_SUSTAINABILITY

Specifications

Description pHrodo™ Deep Red, 250 μL
For Use With (Application) Antibody binding screening
Product Type Labeling Reagent
Quantity 250 μL
Content And Storage Store in refrigerator (2°C to 8°C) and protect from light
Detection Method Fluorescence
Excitation/Emission 640/655 nm
Product Line pHrodo
Label or Dye pHrodo Deep Red
Color Deep Red
Labeling Target IgG
Species Human
Label Type pHrodo™ (pH sensitive)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.