missing translation for 'onlineSavingsMsg'
Learn More

Zeolite ferrierite, ammonium, Thermo Scientific Chemicals

Product Code. 15482527
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Zeolite ferrierite, ammonium is used as a commercial filter and ion-exchange bed. Further, it acts as a catalyst for the acid-catalyzed isomerization of n-butenes to isobutene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Zeolite ferrierite is used as commercial filters, ion-exchange beds, and as a catalyst for the acid-catalyzed isomerization of n-butenes to isobutene
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1318-02-1
Molecular Formula Al2O5Si
Molecular Weight (g/mol) 162.043
MDL Number MFCD00132601
InChI Key HNPSIPDUKPIQMN-UHFFFAOYSA-N
Synonym kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
PubChem CID 9942228
IUPAC Name dioxosilane;oxo(oxoalumanyloxy)alumane
SMILES O=[Al]O[Al]=O.O=[Si]=O

Specifications

Color White
Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water.
Chemical Name or Material Zeolite ferrierite, ammonium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'