Learn More
Zeolite Y, hydrogen, Thermo Scientific Chemicals
45.02 € - 466.00 €
Chemical Identifiers
CAS | 1318-02-1 |
---|---|
Molecular Formula | Al2O5Si |
Molecular Weight (g/mol) | 162.043 |
MDL Number | MFCD00132601 |
InChI Key | HNPSIPDUKPIQMN-UHFFFAOYSA-N |
Synonym | kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural |
PubChem CID | 9942228 |
IUPAC Name | dioxosilane;oxo(oxoalumanyloxy)alumane |
SMILES | O=[Al]O[Al]=O.O=[Si]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15442497
|
Thermo Scientific Alfa Aesar
045872.14 |
25 g |
45.02 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15452497
|
Thermo Scientific Alfa Aesar
045872.22 |
100 g |
92.46 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15462497
|
Thermo Scientific Alfa Aesar
045872.36 |
500 g |
466.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other productsZeolite Y is used as a cracking catalyst. In the petrochemical industry, it is used in acidic form in order to increase the yield of gasoline and diesel fuel from crude oil feedstock by cracking heavy paraffins into gasoline grade naphthas. Further, it is employed in the hydrocracking units as a platinum/palladium support to increase the aromatic content of reformulated refinery products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products
Chemical Identifiers
1318-02-1 | |
162.043 | |
HNPSIPDUKPIQMN-UHFFFAOYSA-N | |
9942228 | |
O=[Al]O[Al]=O.O=[Si]=O |
Al2O5Si | |
MFCD00132601 | |
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural | |
dioxosilane;oxo(oxoalumanyloxy)alumane |
Specifications
1318-02-1 | |
Powder | |
MFCD00132601 | |
Insoluble in water. | |
O=[Al]O[Al]=O.O=[Si]=O | |
162.043 | |
Zeolite Y, hydrogen |
White | |
Al2O5Si | |
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural | |
HNPSIPDUKPIQMN-UHFFFAOYSA-N | |
dioxosilane;oxo(oxoalumanyloxy)alumane | |
9942228 |
Safety and Handling
missing translation for 'einecsNumber' : 215-283-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only