Learn More
ZVG™ ZetClean™ Wiper Bowl™ Multitex™ Wipe Dispenser Buckets
ZetClean™ Wiper Bowl™ Multitex™ moist, perforated cleaning wipes are tear-resistant, lint-free, and come in a convenient dispenser bucket. These wipes boast an orange scent.
Brand:  ZVG™ 50132-000
Product Code. 17833675
  • 86.95 € / Pack of 6
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The moist Multitex™ wipes from the zetClean™ product range, soaked in a special cleaning solution and highly tear-resistant, are indispensable for cleanliness in workshops and machine parks. They are particularly ideal for workplaces where immediate access to water for cleaning is not available. The soft, tear-resistant viscose wipes with textile-like characteristics are enriched with Vitamin E and aloe vera, making them especially gentle on the skin. They are suitable for removing oils, fats, adhesives, tar, grease, and more.
  • Viscose, nonwoven wipes with textile-like characteristics
  • Suitable for wet and dry cleaning
  • Robust and lint-free
  • Tear-resistant
  • Soaked in a gentle cleaning solution
  • Pleasant fresh orange scent
  • Easy tear-off perforation for convenient use
  • Resealable dispenser bucket
  • 72 wipes per bucket
  • Particularly skin-friendly
Specifications
Specifications
Wiper
25 cm
25 cm
Blue
Non-sterile
9.8 in.
9.8 in.
Polypropylene, Viscose, Cotton
Single-use