missing translation for 'onlineSavingsMsg'
Learn More

ZVG™ ZetClean™ Wiper Bowl™ Polytex™ Wipe Dispenser Buckets

ZetClean™ Wiper Bowl™ Polytex™ moist cleaning wipes are durable, lint-free wipes in a dispenser bucket. These wipes boast a clean, orange scent.

Brand:  ZVG™ 50130-001

Product Code. 17883675

  • 75.35 € / Pack of 6
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The moist Polytex™ wipes from the zetClean™ product range are indispensable for cleanliness in any workplace, especially in workplaces where water for hand cleaning is not readily available. The Polytex™ wipes, soaked in gentle cleaning agents, are suitable for removing fats, adhesives, ink, wax, coal, graphite, grass stains, and even stubborn contaminants, such as tar or grease. Thanks to Vitamin E and aloe vera, they are particularly skin-friendly. The cleaning wipes are packed in a convenient Wiper Bowl™ dispenser bucket, ensuring that the moist cleaning agents remain effective for a long time and do not dry out.

  • High-quality wipes for versatile use
  • Highly durable and lint-free
  • 100% polypropylene
  • Soaked in gentle cleaning agents
  • Pleasant fresh orange scent
  • Easy tear-off perforation for convenient use
  • Resealable dispenser bucket
  • Particularly skin-friendly thanks to Vitamin E and Aloe Vera
Specifications

Specifications

Wiper
25 cm
25 cm
Turquoise
Non-sterile
9.8 in.
9.8 in.
Polypropylene
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.