missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Newtex™ ZetexPlus™ 200 Series Heat Resistant Gloves
Shop All Cole-Parmer Products
Click to view available options
Length (English):
14 in.
Description
- Vermiculite coating evenly dissipates heat across the surface of the glove
- Coating significantly increases temperature rating and abrasion resistance
- Premium wool liner
- Double stitched and reinforced
- Inorganic fibers will not produce hazardous outgassing
Specifications
Specifications
Coating Material | Vermiculite |
Color | Tan |
Material | ZetexPlus Fiberglass Fabric |
Size | Universal |
Temperature (English) Max. | 1500°F (Extended), 2000°F (Intermittent) |
Temperature (Metric) Max | 815°C (Extended), 1095°C (Intermittent) |
For Use With (Application) | Kiln/Furnace Operations, Heat Processing Facilities, and High Temperature Industrial Environments |
Glove Cuff Style | Open |
Liner Material | Wool |
Series | ZetexPlus™ 200 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Newtex™ ZetexPlus™ 200 Series Heat Resistant Gloves >
Spot an opportunity for improvement?Share a Content Correction