missing translation for 'onlineSavingsMsg'
Learn More

ZVG™ zetMedica™ Eco-friendly Medical Rolls

These medical rolls made of recycled material are perforated and feature dot-to-dot embossing, allowing for sustainable use in medical applications.

Brand:  ZVG™ 12315-01

 View more versions of this product

Product Code. 17813525

  • 126.00 € / 6 rolls

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Effective in quickly preparing the premises in the care area for the next patient
  • Tissue is soft and highly absorbent
  • Material does not stick to the skin, and immediately absorbs moisture and grease
  • Rolls are economically usable thanks to their perforation and always remain clean at the tear-off edge
  • Suitable doctor's roll holder; can be mounted either on the wall or directly on the bed
  • Made of recycled material
  • Contains approximately 260 tear-off sheets per roll
  • With point-to-point embossing

In a novel manufacturing process, the individual layers of the cellulose cloth are bonded together at many points across the entire surface. This creates a strong bond between the layers, making them highly tear-resistant. This process combines the advantages of a thick wiping cloth (stability) with those of a multi-layered cloth (high absorbency).

Specifications

Specifications

Roll Paper
Paper
Single-use
2-ply
10.94 kg
White
Non-sterile
100 m
6 Rolls
50 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.