missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEPperCHIP™ Zika Virus Proteome Microarray
Zika Virus Proteome Microarray with 3419 virus specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.
Brand: PEPperCHIP™ SPC.046.001
This item is not returnable.
View return policy
Description
- Microarray Content: Zika virus genome polyprotein cleaved into 13 chains: protein C, protein prM, peptide pr, small envelope protein M, envelope protein E, non-structural protein 1, non-structural protein 2A, serine protease subunit NS2B, serine protease subunit NS3, non-structural protein 4A, peptide 2K, non-structural protein 4B, RNA-directed RNA polymerase NS5
- Peptide Length/Overlap: 15 aa/ 14 aa
- Number of Peptides: 3419 printed in duplicate and HA, polio and c-Myc control peptides
Specifications
PEGMA/PMMA | |
1 Slide | |
1 mm | |
Transparent | |
Individual barcode for each microarray | |
High-density Peptide Microarray | |
Non-sterile |
1 array copy per microarray | |
Combinatorial peptide synthesis by amino acid laser printing | |
ISO 9001:2015 | |
RUO | |
Microscopic Glass | |
Antibody validation, epitope mapping, serum screening, biomarker discovery | |
75.4 x 25 mm |