Learn More
Zinc 2-ethylhexanoate, ca 80% in mineral spirits (17-19% Zn), Thermo Scientific Chemicals
38.49 € - 475.37 €
Chemical Identifiers
CAS | 136-53-8 |
---|---|
Molecular Formula | C16H30O4Zn |
Molecular Weight (g/mol) | 351.792 |
MDL Number | MFCD00053316 |
InChI Key | IFNXAMCERSVZCV-UHFFFAOYSA-L |
Synonym | zinc 2-ethylhexanoate, zinc bis 2-ethylhexanoate, zinc 2-ethylhexoate, hexanoic acid, 2-ethyl-, zinc salt, hexanoic acid, 2-ethyl-, zinc salt 2:1, zinc ethylhexanoate, hexanoic acid, 2-ethyl-, zinc salt, basic, zinc 2-ethylhexanoate 100g, zinc 2+ ion bis 2-ethylhexanoate |
PubChem CID | 61083 |
IUPAC Name | zinc;2-ethylhexanoate |
SMILES | CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zn+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11368378
|
Thermo Scientific Alfa Aesar
039561.22 |
100 g |
38.49 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11420231
|
Thermo Scientific Alfa Aesar
039561.36 |
500 g |
116.58 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430231
|
Thermo Scientific Alfa Aesar
039561.0E |
2500 g |
475.37 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is applied as a catalyst for condensation-cure silicones and as a vulcanization activator in industries. It is also employed in PVC heat stabilizer, polyurethane catalyst and lubricant additive. They are used as catalysts in paints and coatings, in polyurethanes, greases, and as wetting agents for pigments. Also used in the formulation of lubricants and greases, as hardener for epoxy resins, to make rubber and plastic products, and for refining crude oil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
136-53-8 | |
351.792 | |
IFNXAMCERSVZCV-UHFFFAOYSA-L | |
61083 | |
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zn+2] |
C16H30O4Zn | |
MFCD00053316 | |
zinc 2-ethylhexanoate, zinc bis 2-ethylhexanoate, zinc 2-ethylhexoate, hexanoic acid, 2-ethyl-, zinc salt, hexanoic acid, 2-ethyl-, zinc salt 2:1, zinc ethylhexanoate, hexanoic acid, 2-ethyl-, zinc salt, basic, zinc 2-ethylhexanoate 100g, zinc 2+ ion bis 2-ethylhexanoate | |
zinc;2-ethylhexanoate |
Specifications
136-53-8 | |
1.07 g/mL | |
MFCD00053316 | |
zinc 2-ethylhexanoate, zinc bis 2-ethylhexanoate, zinc 2-ethylhexoate, hexanoic acid, 2-ethyl-, zinc salt, hexanoic acid, 2-ethyl-, zinc salt 2:1, zinc ethylhexanoate, hexanoic acid, 2-ethyl-, zinc salt, basic, zinc 2-ethylhexanoate 100g, zinc 2+ ion bis 2-ethylhexanoate | |
IFNXAMCERSVZCV-UHFFFAOYSA-L | |
zinc;2-ethylhexanoate | |
61083 | |
Liquid |
Colorless | |
C16H30O4Zn | |
UN1268 | |
Not miscible or difficult to mix in water. | |
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zn+2] | |
351.792 | |
351.79 | |
Zinc 2-ethylhexanoate |
Safety and Handling
GHS H Statement
H340-H350-H372-H304-H361-H226
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
Flammable liquid and vapor.
P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280i-P281-P301+P310-P303+P361+P353-P305+P351+P338-P308+P313-P331-P370+P378q-P501c
H226-H304-H319-H340-H350-H361d-H372
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM DISTILLATES, N.O.S.
missing translation for 'einecsNumber' : 205-251-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Zinc 2-ethylhexanoate, ca 80% in mineral spirits
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Aspiration toxicity Category 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H319-Causes serious eye irritation.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P331-Do NOT induce vomiting.
Supplemental information
- MIXTURE LIST-Contains : Zinc 2-ethylhexoate,Stoddard solvent
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.