missing translation for 'onlineSavingsMsg'
Learn More

Zinc 2-ethylhexanoate, Zn ≈ 20%, cont. 1% diethylene glycolmonomethyl ether, Thermo Scientific Chemicals

55.10 € - 208.00 €

Chemical Identifiers

CAS 136-53-8
Molecular Formula C16H30O4Zn
Molecular Weight (g/mol) 351.792
MDL Number MFCD00053316
InChI Key IFNXAMCERSVZCV-UHFFFAOYSA-L
Synonym zinc 2-ethylhexanoate, zinc bis 2-ethylhexanoate, zinc 2-ethylhexoate, hexanoic acid, 2-ethyl-, zinc salt, hexanoic acid, 2-ethyl-, zinc salt 2:1, zinc ethylhexanoate, hexanoic acid, 2-ethyl-, zinc salt, basic, zinc 2-ethylhexanoate 100g, zinc 2+ ion bis 2-ethylhexanoate
PubChem CID 61083
IUPAC Name zinc;2-ethylhexanoate
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zn+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11358378
View Documents
Thermo Scientific Alfa Aesar
039560.22
100 g
55.10 €
100g
Please to purchase this item. Need a web account? Register with us today!
11348378
View Documents
Thermo Scientific Alfa Aesar
039560.36
500 g
208.00 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zinc 2-ethylhexanoate is widely used in polyvinylchloride heat stabilizer, polyurethane catalyst and lubricant additive. It is an auxiliary drier used as pigments dispersing agent and prevents the paint film from webbing and frosting effects. It is involved in the preparation of highly porous dentritic zinc oxide nanostructures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

136-53-8
351.792
IFNXAMCERSVZCV-UHFFFAOYSA-L
61083
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zn+2]
C16H30O4Zn
MFCD00053316
zinc 2-ethylhexanoate, zinc bis 2-ethylhexanoate, zinc 2-ethylhexoate, hexanoic acid, 2-ethyl-, zinc salt, hexanoic acid, 2-ethyl-, zinc salt 2:1, zinc ethylhexanoate, hexanoic acid, 2-ethyl-, zinc salt, basic, zinc 2-ethylhexanoate 100g, zinc 2+ ion bis 2-ethylhexanoate
zinc;2-ethylhexanoate

Especificaciones

136-53-8
MFCD00053316
Immiscible with cold water.
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zn+2]
351.792
351.79
Zinc 2-ethylhexanoate
C16H30O4Zn
zinc 2-ethylhexanoate, zinc bis 2-ethylhexanoate, zinc 2-ethylhexoate, hexanoic acid, 2-ethyl-, zinc salt, hexanoic acid, 2-ethyl-, zinc salt 2:1, zinc ethylhexanoate, hexanoic acid, 2-ethyl-, zinc salt, basic, zinc 2-ethylhexanoate 100g, zinc 2+ ion bis 2-ethylhexanoate
IFNXAMCERSVZCV-UHFFFAOYSA-L
zinc;2-ethylhexanoate
61083
Liquid
Videos
Seguridad y manipulación

Seguridad y manipulación

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.

P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c

H319-H361d

missing translation for 'einecsNumber' : 205-251-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documentos

Documentos

Certificados

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado