missing translation for 'onlineSavingsMsg'
Learn More
Zinc, +99%, mossy, Thermo Scientific Chemicals Product Code.: 10742821

Zinc, +99%, mossy, Thermo Scientific Chemicals

Product Code. 10742821
500 g, Plastic bottle
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10742821

Brand: Thermo Scientific Acros 201455000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Gray
Melting Point 419.0°C
Boiling Point 907.0°C
Density 7.1400g/mL
Assay Percent Range 99+%
Linear Formula Zn
Quantity 500 g
Fieser 01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406
Merck Index 15, 10326
Formula Weight 65.38
Percent Purity ≥99%
Packaging Plastic bottle
Form Flake
Chemical Name or Material Zinc, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469021 Certificate TypeCertificate of Analysis Date20/01/2025 Catalog Number201450010,201455000,201450050
Lot NumberA0460921 Certificate TypeCertificate of Analysis Date14/03/2024 Catalog Number201450010,201455000,201450050
Lot NumberA0400397 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number201450010,201455000,201450050
Lot NumberA0404720 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number201450010,201455000,201450050
Lot NumberA0414004 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number201450010,201455000,201450050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zinc, +99%, mossy, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.