missing translation for 'onlineSavingsMsg'
Learn More

Zinc acetate dihydrate, 98%, extra pure

Product Code. 10581841 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10581841

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5970-45-6 | C4H6O4Zn·2H2O | 219.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H6O4Zn·2H2O
Molecular Weight (g/mol) 219.5
MDL Number MFCD00066961
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Specifications

Melting Point 237.0°C
Color White
Physical Form Crystals
Quantity 1 kg
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula (CH3CO2)2Zn·2H2O
Merck Index 15, 10327
Solubility Information Solubility in water: 430g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 219.5
Percent Purity 98%
Grade Extra Pure
Packaging Plastic bottle
Specific Gravity 1.84
Density 1.8400g/mL
Chemical Name or Material Zinc acetate dihydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc acetate dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.