missing translation for 'onlineSavingsMsg'
Learn More

Zinc acrylate, Thermo Scientific Chemicals

Product Code. 11378928
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

The primary use of Zinc Acrylate is in the production of golf balls. Other uses are primarily in the production of specialty rubber products such as belts and hoses for automobiles. Also used as a tissue adhesive used for adherence of tissue to tissue or tissue to non-tissue for prostheses in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14643-87-9
Molecular Formula C6H6O4Zn
Molecular Weight (g/mol) 207.49
MDL Number MFCD00067208
InChI Key XKMZOFXGLBYJLS-UHFFFAOYSA-L
Synonym zinc acrylate, zinc diacrylate, 2-propenoic acid, zinc salt, acrylic acid, zinc salt, 2-propenoic acid, zinc salt 2:1, zinc prop-2-enoate, 79-10-7 parent, acmc-20aj8t, dsstox_cid_7768, dsstox_rid_78559
PubChem CID 159747
IUPAC Name zinc;prop-2-enoate
SMILES [Zn++].[O-]C(=O)C=C.[O-]C(=O)C=C

Specifications

Melting Point ∼250°C
Density 1.6 g/mL
Quantity 250 g
UN Number UN3077
Solubility Information Soluble completely in acrylic acid and water (as a salt). Insoluble as a polymer.
Formula Weight 207.5
Physical Form Powder
Chemical Name or Material Zinc acrylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.