missing translation for 'onlineSavingsMsg'
Learn More

Zinc bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Product Code. 15462787
Click to view available options
Quantity:
250 mg
1 g
5 g

Zinc bis(trifluoromethylsulfonyl)imide is used as a Catalyst for Enantioselective Friedel-Crafts alkylation of pyrroles with β-CF3 acrylates, Stereoselective Diels-Alder reaction, Tandem cyclopropane ring-opening / Conia-ene cyclization Aromatic nitration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 168106-25-0
Molecular Formula C4F12N2O8S4Zn
Molecular Weight (g/mol) 625.651
MDL Number MFCD16621474
InChI Key QEORIOGPVTWFMH-UHFFFAOYSA-N
Synonym Zinc triflimide; Zinc trifluoromethanesulfonimide
PubChem CID 131849436
IUPAC Name zinc;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Zn+2]

Specifications

Melting Point 143°C to 147°C
Color White
Flash Point >110°C (230°F)
Linear Formula [(CF3SO2)2N]2Zn
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information It is soluble in CH2Cl2 and H2O.
Formula Weight 625.65
Physical Form Solid
Chemical Name or Material Zinc bis (trifluoromethylsulfonyl)imide
compliance-icons
Product Identifier
  • Zinc bis(trifluoromethylsulfonyl)imide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Zinc bis(trifluoromethylsulfonyl)imide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.