Learn More
Zinc bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
Description
Zinc bis(trifluoromethylsulfonyl)imide is used as a Catalyst for Enantioselective Friedel-Crafts alkylation of pyrroles with β-CF3 acrylates, Stereoselective Diels-Alder reaction, Tandem cyclopropane ring-opening / Conia-ene cyclization Aromatic nitration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 143°C to 147°C |
Color | White |
Flash Point | >110°C (230°F) |
Linear Formula | [(CF3SO2)2N]2Zn |
Quantity | 250 mg |
Sensitivity | Moisture sensitive |
Solubility Information | It is soluble in CH2Cl2 and H2O. |
Formula Weight | 625.65 |
Physical Form | Solid |
Chemical Name or Material | Zinc bis (trifluoromethylsulfonyl)imide |
Safety and Handling
- Zinc bis(trifluoromethylsulfonyl)imide
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Zinc bis(trifluoromethylsulfonyl)imide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.