missing translation for 'onlineSavingsMsg'
Learn More

Zinc Chloride, Reagent grade, ≥98%, Honeywell™

Product Code. 15631820
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl

Specifications

Melting Point 293°C
Color White
Physical Form Solid
Quantity 1 kg
Linear Formula ZnCl2
UN Number UN2331
Formula Weight 136.3g/mol
Percent Purity ≥98%
Grade Reagent
Packaging Poly Bottle
Vapor Pressure 1mmHg (428°C)
Density 2.93g/cu.cm
Chemical Name or Material Zinc Chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.