missing translation for 'onlineSavingsMsg'
Learn More

Zinc chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

117.59 € - 366.83 €

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11341299
View Documents
Thermo Scientific Alfa Aesar
013694.09
10 g
117.59 €
10g
Please to purchase this item. Need a web account? Register with us today!
11351299
View Documents
Thermo Scientific Alfa Aesar
013694.18
50 g
366.83 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zinc(II) chloride is used as a catalyst, as a dehydrating and condensing agent, soldering flux and metal etchant. It is also employed in preserving anatomical specimens, wood preservatives, deodorant, disinfecting and embalming materials. It is employed as a mordant in printing and dyeing materials, and in the vulcanizing processes of fiber and rubber. It is useful as an electrolyte in dry cell batteries, in metal industry, in galvanizing iron and as an electrolyte for electroplating. It is useful in an organic synthesis, for example in the Friedel-crafts acylation, Fisher indole synthesis, Lucas reagent (zinc chloride with HCl), activation of allylic/benzylic halides towards reaction with olefins or with sodium cyanoborohydride catalyzed reduction of halides, organozinc reagents for use in Negishi coupling, and aldol condensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7646-85-7
136.28
JIAARYAFYJHUJI-UHFFFAOYSA-L
5727
dichlorozinc
Cl2Zn
MFCD00011295
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
CHEBI:49976
Cl[Zn]Cl

Specifications

732°C
7646-85-7
Beads
Cl2Zn
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
JIAARYAFYJHUJI-UHFFFAOYSA-L
dichlorozinc
5727
136.29
Hygroscopic
2.91
290°C
White
99.99% metal basis
MFCD00011295
Freely soluble in water and dilute hydrochloric acid. Soluble in alcohol,glycerol,ether and acetone.
Cl[Zn]Cl
136.28
CHEBI:49976
Odorless
1hPa (1mm Hg) at 20°C
Zinc chloride, ultra dry
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III

missing translation for 'einecsNumber' : 231-592-0

missing translation for 'rtecsNumber' : ZH1400000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only