missing translation for 'onlineSavingsMsg'
Learn More
Zinc, granular, 30 mesh, Thermo Scientific Chemicals Product Code.: 10276800

Zinc, granular, 30 mesh, Thermo Scientific Chemicals

Product Code. 10276800
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10276800

Brand: Thermo Scientific Acros 222615000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Gray
Melting Point 420.0°C
Boiling Point 907.0°C
Density 7.1400g/mL
Linear Formula Zn
Quantity 500 g
Fieser 01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406
Merck Index 15, 10326
Formula Weight 65.38
Percent Purity ≥99.7%
Packaging Plastic bottle
Form Fine Granular Powder
Chemical Name or Material Zinc, 30 mesh, p.a.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463746 Certificate TypeCertificate of Analysis Date26/06/2024 Catalog Number222611000,222615000,222610025
Lot NumberA0457319 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number222611000,222615000,222610025
Lot NumberA0402241 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number222611000,222615000,222610025
Lot NumberA0415300 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number222611000,222615000,222610025
Lot NumberA0445092 Certificate TypeCertificate of Analysis Date19/08/2022 Catalog Number222611000,222615000,222610025
compliance-icons compliance-icons
Product Identifier
  • Zinc, granular, 20 mesh
Signal Word
  • Danger
Hazard Category
  • Pyrophoric solid Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use sand to extinguish.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zinc, granular, 30 mesh, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.