missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Iodide, Extra Pure, SLR, Fisher Chemical
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Specifications
Specifications
Melting Point | 446°C |
CAS | 10139-47-6 |
Color | Cream |
pH | 5.6 |
Physical Form | Powder/Solid |
Quantity | 100 g |
Molecular Formula | I2Zn |
MDL Number | 11299 |
Formula Weight | 319.19g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Zinc iodide
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'