missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc neodecanoate, Zn 17.9-18.2%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
5 kg
Description
Zinc neodecanoate is used as a catalyst, lubricant additive and drier. Further it is used in the production of rubber and plastic products. In addition it serves as a polyurethane catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Brown |
Flash Point | 102°C (216°F) |
Quantity | 250 g |
Solubility Information | Immiscible with water. |
Formula Weight | 407.89 |
Physical Form | Liquid |
Chemical Name or Material | Zinc neodecanoate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'