missing translation for 'onlineSavingsMsg'
Learn More

Zinc neodecanoate, Zn 17.9-18.2%, Thermo Scientific Chemicals

Artikelnummer. 11399677
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
1 kg
5 kg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Zinc neodecanoate is used as a catalyst, lubricant additive and drier. Further it is used in the production of rubber and plastic products. In addition it serves as a polyurethane catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 27253-29-8
Molecular Formula C20H40O4Zn
Molecular Weight (g/mol) 409.916
MDL Number MFCD00014384
InChI Key VADFQXONMAETJP-UHFFFAOYSA-N
Synonym zinc neodecanoate, zinc neodecanoate, zn, 7,7-dimethyloctanoyl oxy zincio 7,7-dimethyloctanoate
PubChem CID 117064711
IUPAC Name 7,7-dimethyloctanoic acid;zinc
SMILES CC(C)(C)CCCCCC(=O)O.CC(C)(C)CCCCCC(=O)O.[Zn]

Spezifikation

Color Brown
Flash Point 102°C (216°F)
Quantity 5 kg
Solubility Information Immiscible with water.
Formula Weight 407.89
Physical Form Liquid
Chemical Name or Material Zinc neodecanoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'