missing translation for 'onlineSavingsMsg'
Learn More

Zinc nitrate hexahydrate, 98%, Thermo Scientific Chemicals

35.07 € - 186.93 €

Specifications

Melting Point ca 36°C
CAS 10196-18-6
Physical Form Solid
Molecular Formula N{2}O{6}Zn^0.6 H{2}O
Percent Purity 98%
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15493127
View Documents
Thermo Scientific Alfa Aesar
A16282.22
100 g
35.07 €
100g
Please to purchase this item. Need a web account? Register with us today!
15403137
View Documents
Thermo Scientific Alfa Aesar
A16282.36
500 g
57.39 €
500g
Please to purchase this item. Need a web account? Register with us today!
15483127
View Documents
Thermo Scientific Alfa Aesar
A16282.0E
2500 g
186.93 €
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

ca 36°C
Solid
98%
10196-18-6
N{2}O{6}Zn^0.6 H{2}O
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Zinc nitrate hexahydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Zinc nitrate hexahydrate

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.