missing translation for 'onlineSavingsMsg'
Learn More

Zinc nitrate hexahydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Product Code. 11326158
Click to view available options
Quantity:
50 g
250 g

Zinc nitrate hexahydrate is used as an analytical reagent, catalyst and as a mordant in dyeing. It finds application in electro galvanizing, rubber industry and in the water treatment industry. It is widely used as the precursor for the synthesis of zinc oxide. It is employed in the preparation of the steel phosphating agent. In the field of agriculture, it is used for nourishing and protecting fruit trees from acid rain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10196-18-6
Molecular Formula H12N2O12Zn
Molecular Weight (g/mol) 297.48
MDL Number MFCD00149889
InChI Key JGPSMWXKRPZZRG-UHFFFAOYSA-N
Synonym zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
PubChem CID 15865313
SMILES O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point ∼36°C
Color White
Physical Form Crystalline Aggregates
Quantity 250 g
Linear Formula Zn(NO3)2·6H2O
UN Number UN1514
Merck Index 14,10144
Solubility Information Soluble in water and alcohol.
IUPAC Name zinc(2+) hexahydrate dinitrate
Formula Weight 297.47 (189.38 Anhydrous)
Percent Purity 99.998%
Grade Puratronic™
Odor Odorless
Sensitivity Hygroscopic
Density 2.07 g/mL
Assay (metals basis)
Chemical Name or Material Zinc nitrate hexahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.