missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Nitrate Hexahydrate, purum p.a., ≥99% (KT), Honeywell™
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Melting Point | 38.9°C |
Color | Colorless |
Quantity | 500 g |
Linear Formula | Zn(NO3)2 · 6H2O |
UN Number | UN1514 |
IUPAC Name | zinc(2+) hexahydrate dinitrate |
Formula Weight | 297.49g/mol |
Percent Purity | ≥99% (KT) |
Odor | Weak |
Packaging | Poly Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'