Learn More
Zinc phthalocyanine, 95%, Thermo Scientific Chemicals
45.23 € - 475.37 €
Chemical Identifiers
CAS | 14320-04-8 |
---|---|
Molecular Formula | C32H16N8Zn |
Molecular Weight (g/mol) | 577.916 |
MDL Number | MFCD00041955 |
InChI Key | PODBBOVVOGJETB-UHFFFAOYSA-N |
Synonym | zinc phthalocyanine, znpc, phthalocyanine zinc, zinc ii phthalocyanine, phthalocyanine, zinc, zinc phthalocyanine, dye content 97 % |
PubChem CID | 2735172 |
ChEBI | CHEBI:51218 |
SMILES | C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Zn+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11398358
|
Thermo Scientific Alfa Aesar
039553.04 |
2 g |
45.23 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11388358
|
Thermo Scientific Alfa Aesar
039553.09 |
10 g |
137.69 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11378358
|
Thermo Scientific Alfa Aesar
039553.18 |
50 g |
475.37 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zinc phthalocyanine finds general applications in industry as catalysts, photoconductors and in biomedical field for photodynamic therapy, PDT. ZnPc loaded PLGA nanoparticles is a promising drug delivery system for photodynamic therapy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14320-04-8 | |
577.916 | |
PODBBOVVOGJETB-UHFFFAOYSA-N | |
2735172 | |
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Zn+2] |
C32H16N8Zn | |
MFCD00041955 | |
zinc phthalocyanine, znpc, phthalocyanine zinc, zinc ii phthalocyanine, phthalocyanine, zinc, zinc phthalocyanine, dye content 97 % | |
CHEBI:51218 |
Specifications
14320-04-8 | |
Odorless | |
MFCD00041955 | |
Insoluble in water. | |
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Zn+2] | |
2735172 | |
577.91 | |
Powder |
Purple | |
C32H16N8Zn | |
zinc phthalocyanine, znpc, phthalocyanine zinc, zinc ii phthalocyanine, phthalocyanine, zinc, zinc phthalocyanine, dye content 97 % | |
PODBBOVVOGJETB-UHFFFAOYSA-N | |
577.916 | |
CHEBI:51218 | |
95% | |
Zinc phthalocyanine |
Safety and Handling
missing translation for 'einecsNumber' : 238-262-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.