missing translation for 'onlineSavingsMsg'
Learn More
Zinc shot, 1-5mm (0.04-0.2in), 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11326857

Zinc shot, 1-5mm (0.04-0.2in), 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11326857
250 g
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11326857

Brand: Thermo Scientific Alfa Aesar 010759.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Gray
Appearance Gray shot
Density 7.14g/cm3
Assay Percent Range 99.999% (Metals basis)
Quantity 250 g
Formula Weight 65.3
Odor Odorless
Form Shot
Chemical Name or Material Zinc, 1-5mm (0.04-0.2 in.)

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ28H013 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010759.18,010759.30,010759.A1
Lot NumberY04H018 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010759.18,010759.30,010759.A1
Lot NumberE30M13 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number010759.18,010759.30,010759.A1
Lot NumberM16G028 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010758.18,010759.30,010758.30
Lot NumberX14H031 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010759.18,010759.30,010759.A1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zinc shot, 1-5mm (0.04-0.2in), 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.