missing translation for 'onlineSavingsMsg'
Learn More

Zinc Solution for ICP, 1g/L, HNO3 Matrix, Fisher Chemical™

Product Code. 12996234
Click to view available options
Quantity:
100 mL
250 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 1g/L
Quantity 500 mL
Solution Contains HNO3
Product Identifier
  • Single-element Standard Solution for ICP. Zinc (Zn) 1000mg/l in HNO3 2%
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'