missing translation for 'onlineSavingsMsg'
Learn More

Zinc standard solution, for AAS, 1 mg/ml Zn in 2-5% HCl, Thermo Scientific Chemicals

 

Brand:  Thermo Scientific Acros 196465000

52.22 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.



Product Code. 10790891

  • 54.97 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Zinc standard solution
7440-66-6, 7647-01-0, 7732-18-5, 7646-85-7
500 mL
Zn
Liquid
Colorless
[Zn]
65.38
CHEBI:30185
AAS
1 mg/ml Zn in 2% HCl
Plastic bottle
1.01g/mL
MFCD00011291
997 to 1003 μg/mL of Zn (2+) in 2% HCl (20°C)
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
23994
65.38
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Zinc standard solution, 1 mg/ml Zn in 2% HCl

Signal Word
  • Warning

Hazard Category
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H290-May be corrosive to metals.

Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.

SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469036 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number196465000
Lot NumberA0442784 Certificate TypeCertificate of Analysis Date07/12/2023 Catalog Number196465000
Lot NumberA0457804 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog Number196465000
Lot NumberA0399188 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number196465000
Lot NumberA0398936 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number196465000
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zinc standard solution, for AAS, 1 mg/ml Zn in 2-5% HCl, Thermo Scientific Chemicals > 500mL; Plastic bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only