missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc sulfate concentrate, For 1L standard solution, 0.1 M ZnSO4, Honeywell Fluka™
Click to view available options
Quantity:
1 Ea.
6 x 1 Ea.
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 1 Ea. |
Linear Formula | ZnSO4 |
UN Number | 3082 |
Formula Weight | 161.45g/mol |
Packaging | Ampoule |
Chemical Name or Material | Zinc Sulfate Concentrate |
Safety and Handling
Product Identifier
- Zinc sulfate concentrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Zinc sulphate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'