Learn More
Chemsplash Zip Labcoat T5/6 Fabric, Knit Cuff Product Code.: 30058420
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
Chemsplash Zip Labcoat T5/6 Fabric, Knit Cuff
GREENER_CHOICE
Product Code. 30058420
Large
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Product Code. 30058420
Brand: Chemsplash 2649.400.006
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
Add to basket
Lab Coat is a 2/3 length coat with a collar and zip closure and knitted cuffs.
Chemsplash Lab Coat
  • Chemsplash 55GSM Type PB 6B Fabric
  • Zip Fasten Front with Adhesive Flap Cover
  • Low-Lint Knitted Cuffs
  • Fold Over Collar
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting Fabric
  • PFAS Free
  • Conforms to ISO 14644-1:2015 Cleanroom Class 5 - 9
Description: The Chemsplash Lab Coat is made from Chemsplash fabric, which is soft and microporous. It is strong and resistant to chemical splash and dust. This lab coat is a 2/3 length coat with a collar and zip closure, featuring knitted cuffs. It is anti-static to meet with EN1149-5:2008, lint-free, and suitable for cleanroom applications. It conforms to ISO 14644-1:2015 Cleanroom Class 5 - 9. Chemsplash accessories can be used to protect the wearer from the environment or the environment from the wearer. The range has been designed to allow for maximum comfort and wearability.
Greener Choice Claims
  • Environmental benefits include:
    • Recycled Content
Specifications
Product Type Lab Coat
Size Large
Gender Unisex
Material Microporous Composite Fabric
Color White
Closure Type Zipper
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Resistance Liquids
No. of Pockets None
Latex Free Yes
Autoclavable Autoclavable
Cleanroom Class Class 100-100,000 (ISO 5-8)
Product Title
Chemsplash Zip Labcoat T5/6 Fabric, Knit Cuff >