missing translation for 'onlineSavingsMsg'
Learn More

Merck ZipTip™ Pipet Tips

Sample preparation and precise liquid handling in one device

Brand:  Merck ZTC18S096

 View more versions of this product

Product Code. 10300231

  • 640.00 € / Pack of 96
Estimated Shipment: 15-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Sample preparation and precise liquid handling in one device

A 0.6μL bed of C18, C4, or SCX (strong cation exchange) media is fixed at the end of a 10μL pipet tip such that there is no dead volume; concentrated, purified samples are eluted in 1 to 4μL. For applications requiring smaller elution volumes (e.g., <1μL), ZipTip μ-C18 tips feature a microbed format containing 0.2μL of media.

  • Concentrate and purify 1–100μL biological samples
  • Ideal for purifying and concentrating femtomoles to picomoles of peptides, proteins, or oligonucleotides in minutes prior to mass spectrometry, HPLC, capillary electrophoresis, and other analytical techniques

Order Info

Available in two packaging configurations: resealable trays (packs of 8 and 24 tips) and 96-place tip racks (packs of 96 and packs of 960 tips)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

With C18 Media
Filtered
3.1cm
96/Pk.
ZipTip
10 μL
1.25 in.
Specialty, Media-Fixed
Universal Fit
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers