missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck ZipTip™ Pipet Tips
Sample preparation and precise liquid handling in one device
Brand: Merck ZTSCXS008
This item is not returnable.
View return policy
Description
Sample preparation and precise liquid handling in one device
A 0.6μL bed of C18, C4, or SCX (strong cation exchange) media is fixed at the end of a 10μL pipet tip such that there is no dead volume; concentrated, purified samples are eluted in 1 to 4μL. For applications requiring smaller elution volumes (e.g., <1μL), ZipTip μ-C18 tips feature a microbed format containing 0.2μL of media.
- Concentrate and purify 1–100μL biological samples
- Ideal for purifying and concentrating femtomoles to picomoles of peptides, proteins, or oligonucleotides in minutes prior to mass spectrometry, HPLC, capillary electrophoresis, and other analytical techniques
Order Info
Available in two packaging configurations: resealable trays (packs of 8 and 24 tips) and 96-place tip racks (packs of 96 and packs of 960 tips)
Specifications
With Strong Cation Exchange Media | |
1.2 in. | |
Specialty, Media-Fixed | |
Universal Fit | |
Polypropylene |
10 μL | |
3cm | |
8/Pk. | |
ZipTip |