missing translation for 'onlineSavingsMsg'
Learn More

Zirconium boride, 99.5% (metals basis excluding Hf), Thermo Scientific Chemicals

Product Code. 11356508
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Zirconium boride is use to improve resistance in zirconia-based, carbon-bonded refractories in contact with ferrous melts. Used in nuclear applications, aerospace refractory, in cutting tools and to protect thermocouple tubes. It also used in use of its relatively high conductivity, especially for a ceramic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12045-64-6
Molecular Formula B2Zr
Molecular Weight (g/mol) 112.844
MDL Number MFCD00064648
InChI Key VWZIXVXBCBBRGP-UHFFFAOYSA-N
Synonym Zirconium diboride
PubChem CID 9812765
IUPAC Name boron;zirconium
SMILES [B].[B].[Zr]

Specifications

Melting Point 3000°C
Physical Form Powder
Quantity 50 g
UN Number UN3178
Solubility Information Insoluble in water.
Formula Weight 112.84
Percent Purity 99.5%
Packaging Plastic bottle
Chemical Name or Material Zirconium boride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'