missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium carbide, 99.5% (metals basis excluding Hf), Hf< 200ppm, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
Zirconium carbide commercially used in tool bits for cutting tools. It is used as an abrasive, in cladding, in cermets and incandescent filaments. It finds application in the coating of uranium dioxide and thorium dioxide particles of nuclear fuel as well as in thermo-photovoltaic radiators and field emitter tips and arrays. It is involved in the usage as refractory coatings in nuclear reactors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 3540°C |
Color | Gray |
Physical Form | Powder |
Quantity | 50 g |
UN Number | UN3178 |
Solubility Information | Soluble in concentrated sulfuric acid,hydrofluoric acid and nitric acid. Insoluble in water. |
Formula Weight | 103.23 |
Percent Purity | 99.5% |
Odor | Odorless |
Density | 6.73 g/mL |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'