missing translation for 'onlineSavingsMsg'
Learn More

Zirconium carbonate, basic hydrate, Thermo Scientific Chemicals

Product Code. 11349338
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Zirconium Carbonate is used in the manufacturing of paint drier auxiliaries, pigments, water proof.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57219-64-4
Molecular Formula CH2O7Zr2
Molecular Weight (g/mol) 308.47
MDL Number MFCD00172265
InChI Key ACWOHNQUPJAZIT-UHFFFAOYSA-J
Synonym hydrogen carbonate; zirconium 4+, zirconium 4+ ion tetrabicarbonate, zirconium iv carbonatebasic
PubChem CID 22645944
IUPAC Name [({[hydroxy(oxo)zirconio]oxy}carbonyl)oxy]zirconiumoylol
SMILES O[Zr](=O)OC(=O)O[Zr](O)=O

Specifications

Color White
Physical Form Powder
Quantity 500 g
Solubility Information Insoluble in water. Soluble in hot concentrated acids.
Formula Weight 290.45 (Anhydrous)
Density 2.2 g/mL
Chemical Name or Material Zirconium carbonate, Basic hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'